Principaux documents
CLS3953312C
Pyrex® gas-dispersion tube
pore size 40-60 μm
Synonyme(s) :
Corning® 39533
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Corning 39533-12C
Dimension de pores
40-60 μm
Description générale
With 12 mm O.D. fritted cylinder, 250 mm L × 8 mm diam. stem. Used to disperse gas in liquids. Size of bubbles depends on porosity. Size permits insertion through small openings.
Caractéristiques et avantages
  • Pyrex® gas-dispersion tube is made of Type I Class A - Low-Expansion Borosilicate Glass.
  • Coarse with 40-60 μm pore size.
  • May withstand autoclaving.
  • Not designed for vacuum and freezing applications.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.