Principaux documents
BR13946
BRAND® spare burette tubes for compact burette
size 25 mL, BLAUBRAND®, clear borosilicate glass 3.3
Synonyme(s) :
Accessories, BLAUBRAND®, Burettes, Compact Burettes
1 EA
61,90 $
61,90 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
61,90 $
About This Item
Code UNSPSC :
41121808
Nomenclature NACRES :
NB.43
61,90 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
clear borosilicate glass 3.3
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 13946
Longueur
705 mm
Taille
25 mL
Catégories apparentées
Description générale
The BRAND® spare burette tubes for compact burettes are crafted from high-quality Boro 3.3 clear glass, ensuring both durability and visibility for accurate liquid measurement. These spare tubes are specifically designed to be compatible with BLAUBRAND® and SILBERBRAND compact burettes, making them a reliable choice for maintaining your liquid handling systems.
Informations légales
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG