Documentos Principais
Z722839
Hettich® ROTOFIX 32A/Universal 320/320R centrifuge accessories
Rotor, 2 place for microplates, for UNIVERSAL 320
Selecione um tamanho
1 EA
R$ 12.515,00
R$ 12.515,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 12.515,00
About This Item
Código UNSPSC:
41103900
NACRES:
NB.22
R$ 12.515,00
Check Cart for Availability
Características
to hold 2 x microtrite plates
embalagem
pack of 1 ea
fabricante/nome comercial
Hettich 1460
Parâmetros
2,218 max. RCF
técnica(s)
centrifugation: suitable
compatibilidade
for use with centrifuge models Universal 320/320R
Categorias relacionadas
Descrição geral
Carriers not included.
Informações legais
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 2
1 of 2
Questions
  1. We are planning to get Hettich Universal 320R centrifuge with rotor for deep plate. After search website, I am not clear what is difference between Z722855 and Z722839? what is carriers that was not included in Z722855. Thanks
    1 answer
    1. Product Z722839 is a 2-place swing-out rotor for the Hettich Universal 320/320R. The manufacturer’s part number is 1460. This rotor has a maximum capacity of 10 plates.
      Product Z722855 is a 2-place swing-out rotor for the Hettich Rotina 380/380R. The manufacturer’s part number is 1760. This rotor has a maximum capacity of 10 plates.
      There is a slight difference in the maximum RCF, however these are not interchangeable. For the Universal 320/320R the Z722839 would be required.
      See the manufacturer’s images below:
      Helpful?