Documentos Principais
EZCURVP0W
Millipore
EZ-Pak® External Power Supply
for AC Powered Dispenser
About This Item
Código UNSPSC:
41116129
NACRES:
NB.79
Nível de qualidade
fabricante/nome comercial
EZ-Pak®
aplicação(ões)
bioburden testing
food and beverages
compatibilidade
for use with EZ-PAK® Curve
Categorias relacionadas
Descrição geral
EZ-Pak® External Power Supply is an accessory system designed to enable EZ-Pak® Membrane Dispenser to work without a direct power supply. The Lithium Ion Battery upon fully charged can dispense 10,000 membranes. The EZ-Pak® Curve saves considerable amount of time in opening the membranes for easy use and thereby reduces the risk of contamination.
Aplicação
EZ-Pak® External Power Supply is an auxiliary part of EZ-Pak® Curve which can be used during routine filtration process.
Informações legais
EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany