Documentos Principais
Z743014
Julabo® CORIO CD heating circulator with open bath
CD-B17, capacity 13 L (17 L), temperature 20-150 °C, stainless steel bath, AC/DC input 230 V AC, Schuko plug
Selecione um tamanho
1 EA
R$ 15.956,00
R$ 15.956,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 15.956,00
About This Item
Código UNSPSC:
41103706
NACRES:
NB.77
R$ 15.956,00
Check Cart for Availability
Materiais
stainless steel bath
embalagem
pack of 1 ea
fabricante/nome comercial
Julabo 9012417
faixa de medição de temperatura
20-150 °C
capacidade
13 L (17 L)
externo L × C × A
38 cm × 40 cm × 47 cm
abertura L × C × A
30 cm × 18 cm × 20 cm
Descrição geral
The Open Heating Bath Circulators of the new CORIO series feature high-quality bath tanks made of stainless steel and pump connections.
Your advantages
  • Models for internal and external applications
  • Bright, white, easy to read display
  • Very quiet
  • Easy pump change-over between internal and external circulation
  • USB connection
  • High-quality bath tanks made of stainless steel
  • Integrated drain screw (except B5)
Cooling coil and bath cover available as optional accessories
Informações legais
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)