Documentos Principais
Z692255
Superfrost® microscope slides
white end, L × W 76 mm × 26 mm
About This Item
Código UNSPSC:
41100000
embalagem
pkg of 100 ea
C × L
76 mm × 26 mm
Informações legais
Superfrost is a registered trademark of Erie Scientific Co.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 3
1 of 3
vibrant-m
S8902
Slides, microscope
Corning® cover glasses square, No. 1, W x L 18 mm x 18 mm
CLS284518
Corning® cover glasses
Jemma Victoria Walker et al.
Nature communications, 10(1), 3596-3596 (2019-08-11)
Stem cells (SCs) receive inductive cues from the surrounding microenvironment and cells. Limited molecular evidence has connected tissue-specific mesenchymal stem cells (MSCs) with mesenchymal transit amplifying cells (MTACs). Using mouse incisor as the model, we discover a population of MSCs