Documentos Principais
Z682055
Ace rotary evaporator condenser complete "C" assembly
plastic-coated glass, fits Büchi® models R-200/R-205 and R-114/R-144
Selecione um tamanho
1 EA
R$ 12.800,00
R$ 12.800,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 12.800,00
About This Item
Código UNSPSC:
41121800
NACRES:
VB.02
R$ 12.800,00
Check Cart for Availability
Materiais
plastic-coated glass
fabricante/nome comercial
Büchi equivalent, 40646
Categorias relacionadas
Descrição geral
The "C" condenser assembly consists of the main glass outer trap body with a 35/20 inner spherical joint bottom connection that attaches to the receiver flasks.
Features standard top and side flange that connects directly to the Büchi® rotary evaporator. The assembly also includes the glass cold trap inner body, PTFE sealing ring with Viton® O-rings, PTFE lid and hose barbs with GL threaded caps.
Informações legais
Büchi is a registered trademark of Büchi Labortechnik AG
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Escolha uma das versões mais recentes:
Certificados de análise (COA)