Z150355
Aldrich® septum-inlet adapter with stopcock
Plain, joint: ST/NS 24/40, PTFE stopcock
About This Item
Produtos recomendados
Materiais
PTFE stopcock
Junta
joint: ST/NS24/40
diâmetro interno
14 mm
Descrição geral
Requires septum Z564702 or Z564699.
Septum-inlets require septum Z565695 or Z565679 except where noted.
Ligação
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC