Documentos Principais
R245291
4,6-BIS-(4-CHLORO-PHENYL)-2,3-DIPHENYL-PYRANYLIUM, TETRAFLUORO BORATE
AldrichCPR
Selecione um tamanho
25MG
R$ 1.123,00
R$ 1.123,00
Previsão de entrega em28 de maio de 2025
Selecione um tamanho
Alterar visualização
25MG
R$ 1.123,00
About This Item
Fórmula linear:
C29H19BCl2F4O
Peso molecular:
541.185
Número MDL:
R$ 1.123,00
Previsão de entrega em28 de maio de 2025
Outras notas
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Escolha uma das versões mais recentes:
Certificados de análise (COA)