Documentos Principais
BR88650
BRAND® stopcock T-bore with nozzles
for tube I.D. 3 mm
Sinônimo(s):
PTFE, Stopcocks
Selecione um tamanho
1 EA
R$ 674,00
R$ 674,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 674,00
About This Item
Código UNSPSC:
41100000
NACRES:
NB.35
R$ 674,00
Check Cart for Availability
Materiais
PTFE
embalagem
pack of 1 ea
fabricante/nome comercial
BRAND 88650
D.I.
2 mm
Categorias relacionadas
Descrição geral
The BRAND® T-bore stopcock with nozzles is made from high-quality PTFE, providing outstanding chemical resistance for a wide range of laboratory applications. Featuring a valve lever constructed from polypropylene (PP), it ensures smooth operation and control. The stopcock is designed with integrated hose nozzles, facilitating efficient fluid transfer and easy connection to hoses. Its robust construction guarantees reliable performance, making it ideal for handling aggressive chemicals and solvents.
Informações legais
BRAND is a registered trademark of BRAND GMBH + CO KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)