Documentos Principais
BR159600
BRAND® water jet filter pump
polypropylene
Sinônimo(s):
Water jet filter pump
Selecione um tamanho
1 EA
R$ 596,00
R$ 596,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 596,00
About This Item
Código UNSPSC:
41105100
NACRES:
NB.79
R$ 596,00
Check Cart for Availability
Materiais
polypropylene
fabricante/nome comercial
BRAND 159600
Descrição geral
For generating vacuum and extracting liquids and vapors*.
Designed for excellent performance: Very low water consumption, constant ulitmate pressure and high suction capacity.
  • High chemical resistance, fluid path consists of PP, FKM and PTFE
  • Operating temperature up to max. 80°C
  • Integrated non-return valve increases safety
  • Simple operation and easy to clean
  • Detachable vacuum connection
  • Variety of supplied adapters simplify connections to most water sources
  • Optional reducing adapters are available
Includes: Water connection (sleeve nut R 3/4 in., reducing adapter R 1/2 in., and tubing connector (hose nozzle) of 10-12 mm outer-), suction line connection (Detachable hose nozzle of 6-9 mm outer-, with screw cap GL 14).
* If necessary, insert an in-line over-flow vessel, or cold trap.
Características e benefícios
  • Good chemical resistance
  • Integrated non-return valve increases safety
  • Simple operation and easy to clean
Informações legais
BRAND is a registered trademark of BRAND GMBH + CO KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 1
1 of 1
L-Glutamic acid ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
G1251
L-Glutamic acid