Principaux documents
TMO140627
Nunc® Cell Culture Inserts in 24-well Nunclon Delta surface plate
cell culture treated, polycarbonate, sterile, pore size 3.0 μm, case of 48 ea (supplied as 4 x 24 well plates, each containing 12 inserts)
Synonyme(s) :
multidish membrane insert, treated membrane insert
48 EACH
494,00 €
494,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
48 EACH
494,00 €
About This Item
Code UNSPSC :
41122102
Nomenclature NACRES :
NB.13
494,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polycarbonate
Stérilité
sterile
Conditionnement
case of 48 ea (supplied as 4 x 24 well plates, each containing 12 inserts)
Fabricant/nom de marque
Nunc 140627
Dimension de pores
3.0 μm pore size
Type de liaison
cell culture treated
Description générale
Cultivate most cell types without matrix coating when using Thermo Scientific Nunc Polycarbonate Membrane Inserts in Multidishes.
Informations légales
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries