Principaux documents
ZFCBEQXUS
Milli-Q
Country box Milli-Q® EQ 7008/16 US
Available only in USA
ZFCBEQXUS
158,00 €
158,00 €
Date d'expédition estimée le04 mars 2025Détails
Changer de vue
ZFCBEQXUS
158,00 €
About This Item
Code UNSPSC :
41104200
Nomenclature NACRES :
JA.13
158,00 €
Date d'expédition estimée le04 mars 2025Détails
Compatibilité
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
Description générale
The Country box for the Milli-Q® EQ 7008/16 system is an installation accessory that contains a printed quick reference guide, a user manual in local language and in English on a USB key, and a power cord adapted to the local electrical network.
Application
The components of the Country box assist users to quickly access the information they need to correctly use their system and connect to the local power network.
Informations légales
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany