Key Documents
57101-U
Supelco
Visiprep 5-port Flask Vacuum Manifold
DL (Disposable Liner)
Select a Size
1 EA
$3,260.00
$3,260.00
Check Cart for Availability
Request a Bulk OrderRequest a Sample
Select a Size
Change View
1 EA
$3,260.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.21
$3,260.00
Check Cart for Availability
Request a Bulk OrderRequest a Sample
Quality Level
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
General description
The Visiprep 5-Port Flask Vacuum Manifold enables analysts using Supelco solid phase extraction tubes to simultaneously prepare up to 5 samples. Unlike conventional vacuum manifolds, the Visiprep 5-Port Flask Manifold allows users to collect their SPE eluate directly into 50 mL round or flat bottom flasks for direct Rotovap evaporation. The manifold consists of a chemical resistant 5-port cover (DL or standard available), gasket, base, a glass basin, vacuum gauge and bleed valve, 5 flow control valves, 5 replaceable solvent guide needles, and a base plate that supports up to five 50 mL round or flat bottom flasks.
Each port on both the standard and DL Visiprep models are equipped with flow control valves.
Recommended Flasks: Aldrich® single-neck flask, 50 mL, joint: ST/NS 24/40
  • Round Bottom (Cat. No. Z414484)
  • Flat Bottom (Cat. No. Z418773)
Unlike the standard Visiprep model, the Visiprep DL (disposable liner) model virtually eliminates the possibility of cross contamination when consecutive samples are extracted on the same manifold port. Threaded through each of the DL manifold ports is an inexpensive and disposable PTFE liner that runs through the solvent guide directly into the collection vessel (50 mL flask). Because reagents and samples exiting the SPE tube can only come in contact with the internal surface of the disposable liner, the risk of cross contamination when processing multiple SPE samples on the same port is virtually eliminated.
Features and Benefits
Supplied with 100 disposable liners (Part No. 57059).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Visiprep is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Chun-Lin Fan et al.
Journal of AOAC International, 98(1), 130-148 (2015-04-11)
A 3 month study was conducted on the ruggedness of a multiresidue method for accuracy and stability. The results indicate that in terms of Youden pair ratios of 201 pesticide aged tea samples falling approximately within 1.00-1.20 of the ratio