Skip to Content
Merck
All Photos(2)

Key Documents

57103-U

Supelco

Visiprep 5-port Flask Vacuum Manifold

standard

Synonym(s):

SPE Vacuum Manifold Flask Accessory

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,860.00

$2,860.00


Estimated to ship on29 March 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 EA
$2,860.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.21

$2,860.00


Estimated to ship on29 March 2025


Request a Bulk OrderRequest a Sample

Quality Level

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

The Visiprep 5-Port Flask Vacuum Manifold enables analysts using Supelco solid phase extraction tubes to simultaneously prepare up to 5 samples. Unlike conventional vacuum manifolds, the Visiprep 5-Port Flask Manifold allows users to collect their SPE eluate directly into 50 mL round or flat bottom flasks for direct Rotovap evaporation. The manifold consists of a chemical resistant 5-port cover (DL or standard available), gasket, base, a glass basin, vacuum gauge and bleed valve, 5 flow control valves, 5 replaceable solvent guide needles, and a base plate that supports up to five 50 mL round or flat bottom flasks.

Each port on both the standard and DL Visiprep models are equipped with flow control valves.

Recommended Flasks: Aldrich single-neck flask, 50 mL, joint: ST/NS 24/40
  • Round Bottom (Cat. No. Z414484)
  • Flat Bottom (Cat. No. Z418773)

Legal Information

3D Insert is a trademark of 3D Biotek, LLC
Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service