Skip to Content
Merck
All Photos(1)

Key Documents

57029

Supelco

Visiprep Manifold Cover (flow control valves and gasket included)

for use with DL (Disposable Liner), 12-port model

Synonym(s):

Cover with 12 DL flow control valves and gasket

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,100.00

$1,100.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 EA
$1,100.00

About This Item

UNSPSC Code:
12000000

$1,100.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for use with DL (Disposable Liner), 12-port model

Looking for similar products? Visit Product Comparison Guide

Footnote

Included with Visiprep DL 12-port Vacuum Manifold (57044)

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service