Skip to Content
Merck
All Photos(1)

Key Documents

57147-U

Supelco

Visiprep Manifold Replacement Flow Control Valve Stem

for use with standard 12- and 24-port models, pk of 24

Synonym(s):

pk24 Valve Stem for Visiprep/Preppy Vacuum Manifold

Sign Into View Organizational & Contract Pricing

Select a Size

24 EA
$304.00

$304.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
24 EA
$304.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.21

$304.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

packaging

pk of 24

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for use with standard 12- and 24-port models

Looking for similar products? Visit Product Comparison Guide

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service