Skip to Content
Merck
All Photos(1)

Key Documents

YY2004K00

Millipore

ESP valve Spare parts kit

316L stainless steel, for cleaning and protecting ESP valves

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$306.00

$306.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 KIT
$306.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

$306.00


Check Cart for Availability

Request more information

Quality Level

material

316L stainless steel
fluoroelastomer O-ring
plastic caps

packaging

pkg of 1 unit
pkg of 10 Plastic caps
pkg of 3 Viton o-rings
pkg of 3 plug seals
pkg of 5 syringe needles

dilution

(Sanitary sampling)

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for cleaning and protecting ESP valves

shipped in

ambient

General description

ESP valve spare parts kit is an accessory for sanitization and protection of Millipore sanitary sampling valves. The sanitary sampling valve is ideal for sterile sampling of high purity water and other liquids in distribution lines or storage tanks.

Application

ESP valve Spare parts kit is used for on-line sampling and filtration of high purity water and other liquids. It is suitable for the traditional bioburden testing of beer, Dairy, Bottled Water, Pharmaceutical products, Soft drink, Cider, Sport drink, and Wine.

Components

  • Viton O-ring x 3
  • Syringe needle x 5
  • Plug seal x 3
  • Plastic caps x 10

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service