Skip to Content
Merck
All Photos(2)

Key Documents

1.46001

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

ICR with non-lockable lid, plate volume × diam. 30 mL × 90 mm, sterile; γ-irradiated, pkg of 10 plates Triple packed, suitable for air monitoring

Synonym(s):

CASO Agar, Casein Soybean Digest Agar, TSA, Tryptone Soya Agar

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$91.50
120 EA
$552.00

$91.50


Available to ship on01 April 2025Details


Request more information

Select a Size

Change View
20 EA
$91.50
120 EA
$552.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

$91.50


Available to ship on01 April 2025Details


Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate

feature

ready-to-use

packaging

pkg of 10 plates Triple packed
box of 120 ea
box of 20 ea

storage condition

protect from light

plate volume × diam.

30 mL × 90 mm

color

light yellow

pH

7.3 ( in H2O)

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

2-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Agar - ICR in 90 mm settle plates is designed for the enumeration of the total aerobic microbial count in air via active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms. The medium supports the growth of non-fastidious as well as moderately fastidious microorganisms. The formulation of the Soybean Casein-Digest medium contains a combination of casein and soy peptones supplying organic nitrogen, particular amino acids and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium and the medium is supplemented with pyruvate to neutralize potential residues of hydrogen peroxide resulting of the decontamination of isolators.

Application

Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total aerobic microbial count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms

Analysis Note

Tryptic Soy Agar - ICR settle plate, is a non-lockable plate and should be used for aerobic incubation only.

Legal Information

MAS-100 is a registered trademark of MBV AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Related Content

Detect microbial contamination gases in a few easy and automated steps. The MAS-100 Atmos® compressed gas sampler is simple and safe to use, and compliant with 21 CFR Part 11.

Settle plates for isolators and critical cleanrooms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service