Skip to Content
Merck
All Photos(2)

Key Documents

1.46552

Millipore

Tryptic Soy Agar - Ready-to-use Contact Plates

Lecithin, Tween®, ICR plus with lockable lid, plate diam. 55 mm, sterile; γ-irradiated, pkg of 10 plates Triple packed, suitable for air monitoring

Synonym(s):

CASO Agar, Casein Soybean Digest Agar with neutralizer, Soybean Casein Digest Agar, TSA, Tryptic Soy Agar, Tryptic Soy Agar with Lecithin and Tween

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
$98.80
200 PKG
$988.00

$98.80


Available to ship on02 April 2025Details


Request more information

Select a Size

Change View
20 PKG
$98.80
200 PKG
$988.00

About This Item

UNSPSC Code:
41106231
NACRES:
NA.74

$98.80


Available to ship on02 April 2025Details


Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate

feature

ready-to-use

composition

Lecithin
Tween®

packaging

pkg of 10 plates Triple packed
box of 20 ea
box of 200 ea

storage condition

protect from light (Tightly closed. Dry)

plate diam.

55 mm

color

light yellow

pH

7.3 ( in H2O)

application(s)

air monitoring
pharmaceutical

compatibility

stand alone

storage temp.

2-25°C

suitability

bacteria
fungi
molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Contact Agar + LT – ICR+ is designed for the determination of the total microbial aerobic and anaerobic count on dry, sanitized surfaces and personnel in Clean rooms and Isolators. It is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The formulation of the Soybean Casein-Digest medium contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. The medium is supplemented with the neutralizers like lecithin and polysorbate 80 (Tween®) and pyruvate to inactivate potential residues of disinfectants on dry surfaces. In addition, ICRplus plates (plus or “+”) are supplied with a lockable lid for safe transport after sampling without the risk of losing the lid. ICRplus plates are suitable for aerobic and anaerobic incubation by turning the lid into the close position for aerobic incubation and into the vent position for anaerobic incubation.

Application

Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total microbial aerobic count on dry, sanitized surfaces and personnel in Isolators and Clean Rooms

Legal Information

TWEEN is a registered trademark of Croda International PLC

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Related Content

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Questions

  1. How long is recommended that the plates can stay out of refrigeration before they are unusable, before returning to refridgeration?

    1 answer
    1. The product can be used for sampling until the expiry date if stored upright, protected from light and
      properly sealed at +15 °C to +25 °C. Condensation can be prevented by avoiding quick temperature shifts and mechanical stress. The testing procedures as described on the Certificate of Analysis can be started up to the expiry date printed on the label.

      For more information please view the data sheets available for this product:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/445/632/tn4342en-ms.pdf
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/350/078/fl5868en-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service