Skip to Content
Merck
All Photos(1)

Key Documents

Z277142

Sigma-Aldrich

Aldrich® deluxe Luer fitting assortment set

for use with small diameterflexible tubing and miniature valves, includes 188 fittings

Synonym(s):

Luer fitting kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$332.00

$332.00


Available to ship on23 April 2025Details



Select a Size

Change View
1 EA
$332.00

About This Item

UNSPSC Code:
41122000
NACRES:
NB.03

$332.00


Available to ship on23 April 2025Details


feature

includes 188 fittings

Related Categories

General description

Includes 188 assorted nylon Luer fittings and clamps for use with small diameter flexible tubing and miniature valves. Parts are supplied in a compartmentalized PP case.

  • 100 male Luer to 1/16, 3/32, 1/8 and 5/32 in. I.D. tubing connectors
  • 15 three-way female Luer tees
  • 15 female Luer connectors
  • 10 male Luer to 10-32 UNF threaded end connectors
  • 10 male Luer to 1/4-28 UNF threaded end connectors
  • 38 assorted tubing clamps

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Please can this be used for emulsion formulation using microfluidic pumps?

    1 answer
    1. This product is 188 assorted nylon Luer fittings and clamps for use with small-diameter flexible tubing and miniature valves. Specific application information is not determined.

      Helpful?

  2. Can you please provide a list including part specification and quantity for each fitting in this kit? The description provided is not detailed enough

    1 answer
    1. The manufacturer has provided the following details on these components. No additional information is necessary.
      1/16 Male Slip Luer for Lock Ring, Nylon, Natural (15)
      3/32 Male Slip Luer for Lock Ring, Nylon, Natural (15)
      1/8 Male Slip Luer for Lock Ring, Nylon, Natural (15)
      1/16 Male Slip Luer, Nylon, Natural (10)
      3/32 Male Slip Luer, Nylon, Natural (10)
      1/8 Male Slip Luer, Nylon, Natural (10)
      5/32 Male Slip Luer, Nylon, Natural (10)
      10-32 UNF Threaded Male Luer, Nylon, Natural (10)
      1/4-28 UNF Threaded Male Luer, Nylon, Natural (10)
      Female x Female Straight Lug Style Luer Adapter, Nylon, Natural (10)
      Female Tee Lug Style Luer Adapter, Nylon, Natural (10)
      Double Grip Hose Clamp (.256-.311 Hold Range) (8)
      Double Grip Hose Clamp (.402-.465 Hold Range) (6)
      Double Grip Hose Clamp (.516-.594 Hold Range) (7)
      Double Grip Hose Clamp (.713-.811 Hold Range) (6)

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service