Skip to Content
Merck
All Photos(1)

Key Documents

YY20040H0

Millipore

ESP valve

316L stainless steel valve

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,180.00

$1,180.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EA
$1,180.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

$1,180.00


Check Cart for Availability

Request more information

Quality Level

material

316 stainless steel stem(s)
316L stainless steel valve
body (1/4" NPTM Halar)

packaging

pkg of 1 unit

dilution

(Sanitary sampling)

input

sample type soft drink
sample type wine
sample type pharmaceutical(s)
sample type sport drink
sample type cider
sample type bottled water
sample type beverage(s)
sample type food(s)
sample type beer
sample type dairy

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

shipped in

ambient

Related Categories

General description

The ESP Sanitary Sampling Valve is ideal for sterile sampling of high purity water and other liquids in distribution lines or storage tanks. The valve can be sanitized effectively and easily to eliminate residual viable contaminants and prevent additional contaminant growth. The unique construction of the valve allows mid-stream sampling and prevents the accumulation of bacteria or particles inside the sampling probe.
The Luer-slip valve outlet is designed for the direct attachment of a Millipore bacteriological monitor or directing liquid flow into any type of sterile receiver. The 18-gauge blunt-end syringe needles are used for sanitizing the unit and the plastic caps to cover the outlet Luer-slip after sanitization.

Application

The ESP Sanitary Sampling Valve is used for on-line sampling and filtration of high purity water and other liquids. It is suitable for the traditional bioburden testing of beer, Dairy, Bottled water, Pharmaceutical products, Soft drink, Cider, Sport drink, and Wine.

Components

  • 18-gauge blunt-end syringe needles x 2
  • Spare O-ring x 1
  • Plug seal x 1
  • Plastic caps x 2

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service