Skip to Content
Merck
All Photos(5)

Key Documents

SYMBCHI01

Millipore

Steritest® Symbio communication hub holder

For isolators, for use with Steritest® Symbio Pump

Synonym(s):

Hub Mount

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$733.00

$733.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 UNIT
$733.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

$733.00


Check Cart for Availability

Request more information

material

stainless steel (epoxy paint)

Agency

EP (2.6.1)
JP (4.06)
USP (71)

packaging

pkg of 1 unit

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio Pump

shipped in

ambient

General description

The Steritest® Symbio communication hub holder for isolators is an accessory which allows to attach the Steritest® Symbio Pump communication hub under the isolator table: left, right, center. The depth can also be adjusted.

Application

Steritest® Symbio communication hub holder for isolator is an accessory for streamlining your workflow and increasing safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service