Skip to Content
Merck
All Photos(2)

Key Documents

SYMBFSI01

Millipore

Steritest® Symbio Flat Seal

For isolator integration of Steritest® Symbio ISL pump

Synonym(s):

Pump integration gasket

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$162.00

$162.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EA
$162.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

$162.00


Check Cart for Availability

Request more information

material

polyoxymethylene seal

Agency

EP (2.6.1)
JP (4.06)
USP (71)

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio pump

shipped in

ambient

General description

The Steritest® Symbio flat seal is an accessory for the Steritest® Symbio ISL pump. Steritest® Symbio ISL Pumps are used for reliable, faster, and easy sterility testing inside an isolator. The flat seal is used to seal the housing top part and the isolator table to ensure airtightness.

Application

Steritest® Symbio flat seal is an accessory for the Steritest® Symbio ISL pumps for streamlining the workflow and increasing safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service