Skip to Content
Merck
All Photos(1)

Key Documents

SYMBFSW01

Millipore

Steritest® pump footswitch

for use with Steritest® Symbio pump, This accessory streamlines the workflow and increases safety during sterility testing

Synonym(s):

Pump foot controller

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$598.00

$598.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 UNIT
$598.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

$598.00


Check Cart for Availability

Request more information

Agency

EP (2.6.1)
JP (4.06)
USP (71)

packaging

pkg of 1 unit

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest® Symbio pump

shipped in

ambient

General description

Command your Steritest Symbio pump with your feet and free-up your hands. Benefit from 2 operating modes : start and stop, and continuous. The footswitch is connected to the pump through a dedicated port on the communication hub.

Application

The Steritest Footswitch is compatible with Steritest® Symbio pumps and with Steritest® Equinox pumps. The footswitch operates in two modes: start/stop and continuous.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service