SYMBCHB01
Steritest® Symbio Communication Hub
Hub for Power Supply and various accessories
Synonym(s):
Communication Hub, Communication Point, Fuse Box, Power supply
Select a Size
About This Item
Recommended Products
material
316L stainless steel housing (with epoxy paint)
Agency
EP (2.6.1)
JP (4.06)
USP (71)
manufacturer/tradename
Steritest®
input
pharmaceutical(s)
application(s)
pharmaceutical
sterility testing
compatibility
for use with Steritest® Symbio pump
shipped in
ambient
General description
The Steritest® Symbio Communication Hub is designed for complete protection of symbio pump in case of power overload during sterility testing or otherwise. The communication hub is equipped with specific fuses to protect the pump components. There are two of them installed:
- One T 6.3A L 250V fuse at front.
- Two T 3.15A L 250V fuses at rear.
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service