Skip to Content
Merck
All Photos(1)

Key Documents

610018

Avanti

O-rings for Extruder

Avanti Research - A Croda Brand

Synonym(s):

O-rings for Extruder, O-rings for Mini-Extruder (2)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$22.00

$22.00


Check Cart for Availability


Select a Size

Change View
1 EA
$22.00

About This Item

UNSPSC Code:
12352211
NACRES:
NB.17

$22.00


Check Cart for Availability

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

15-25°C

General description

Replacement Part for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research Mini-Extruder.

Biochem/physiol Actions

O-rings for extruder serves to support the internal membrane[1] and prevents leakage from the filter.[2]

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Small-volume extrusion apparatus for preparation of large, unilamellar vesicles
MacDonald RC, et al.
Biochimica et Biophysica Acta - Biomembranes, 1061(2), 297-303 (1991)
Liposome Preparation for the Analysis of Lipid-Receptor Interaction and Efferocytosis
Voss OH, et al.
Current Protocols in Immunology, 120(1), 14-44 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service