Skip to Content
Merck
All Photos(1)

Key Documents

610023

Avanti

Extruder Set Without Holder/Heating Block

Avanti Research - A Croda Brand

Synonym(s):

Extruder Set Without Holder/Heating Block

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,320.00

$1,320.00


Check Cart for Availability


Select a Size

Change View
1 EA
$1,320.00

About This Item

UNSPSC Code:
23151503
NACRES:
NB.51

$1,320.00


Check Cart for Availability

feature

Extruder Set without Block

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

room temp

General description

API 972: Extruder Kit Box (9-3/16" x 7-3/16" x 2-7/8")
The Avanti Mini-Extruder allows researchers and scientists to prepare large, unilamellar vesicles by extrusion (LUVET) in an efficient, rapid manner. See brochure under Downloads for additional information.
Watch the video to learn more about the Avanti Research Mini-Extruder.

Application

Extruder Set without holder/heating block has been used to prepare liposome in the absence of cholesterol.[1] It has also been used to prepare large unilamellar vesicles (liposomes) containing carboxyfluorescein.[2]

Packaging

Mini-Extruder (set): includes instructions, mini-extruder, two 1000μL syringes, 100 pore size (0.1μm) polycarbonate membranes, and 100 filter supports

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 610020Mini-Extruder (only), Avanti Research - A Croda BrandSDS

  • 610005PC Membranes 0.1um, Avanti Research - A Croda BrandSDS

  • 610017Syringe (1000 μL), Avanti Research - A Croda BrandSDS

  • 610014Filter Support, Avanti Research - A Croda BrandSDS

also commonly purchased with this product

replacement part

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro Lipid Transfer Assay
Zhang Y and Wang X
Current Biology (2012)
Liposome disruption assay to examine lytic properties of biomolecules
Jimah JR, et al.
Bio-protocol, 7(15) (2017)

Protocols

Watch the below video for instructions on liposome preparation using the Avanti® Mini-Extruder.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service