Skip to Content
Merck
All Photos(1)

Key Documents

610017

Avanti

Syringe (1000 μL)

Avanti Research - A Croda Brand

Synonym(s):

Extruder Syringe, Gas-tight Syringe, 1000μL

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$230.00

$230.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$230.00

About This Item

UNSPSC Code:
41122000
NACRES:
NA.28

$230.00


Available to ship on14 April 2025Details


Request a Bulk Order

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Replacement Part for Avanti′s Mini-Extruder, products 610000 and 610023. Contains plunger, needle, syringe screw cap, and PTFE seals.

Watch the video to learn more about the Avanti Research Mini-Extruder.

Application

Syringe (1000 μL) has been used:
  • to prepare unilamellar-vesicles
  • to transfer cell membrane solution for the preparation of cell membrane vesicles
  • to transfer lipid stocks in organic solvents for liposome preparation

Packaging

Syringe Box (8-3/8" X 1-5/8" X 1-1/4") (610017-1Ea)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

replacement part

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service