Skip to Content
Merck
All Photos(4)

Key Documents

610020

Avanti

Mini-Extruder (only)

Avanti Research - A Croda Brand

Synonym(s):

Mini-Extruder

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$734.00

$734.00


Available to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$734.00

About This Item

UNSPSC Code:
23151503
NACRES:
NB.51

$734.00


Available to ship on10 April 2025Details


Request a Bulk Order

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

room temp

General description

Replacement Part for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research Mini-Extruder.

Application

Mini-Extruder (only) has been used to perform extrusion (fifteen times) of the multilamellar vesicles (MLVs) suspension, through a polycarbonate filter.[1]

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azalomycin F5a, a polyhydroxy macrolide binding to the polar head of phospholipid and targeting to lipoteichoic acid to kill methicillin-resistant Staphylococcus aureus
Yuan G, et al.
Biomedicine and Pharmacotherapy, 109, 1940-1950 (2019)

Protocols

Watch the below video for instructions on liposome preparation using the Avanti® Mini-Extruder.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service