Skip to Content
Merck
All Photos(1)

Key Documents

Z315028

Hirschmann® pipetus® pipette controller accessories

0.2 μm PTFE membrane filters, 1 each = 5 filters

Synonym(s):

Pipette accessories

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$111.00

$111.00


Available to ship on17 April 2025Details



Select a Size

Change View
1 EA
$111.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

$111.00


Available to ship on17 April 2025Details


manufacturer/tradename

Hirschmann 9903704

packaging

pkg of 1 each = 5 PTFE membrane filters supplied

Looking for similar products? Visit Product Comparison Guide

Application

For all Pipetus models

Legal Information

Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Pipetus is a registered trademark of Hirschmann Laborgeräte GmbH & Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service