No, this product is not rechargeable. This item is a manual pipette filler that does not have suction nor delivery that is motor driven. The suction and delivery are regulated by an internal bellows, which is inflated and deflated by the suction and delivery buttons. Please see the link to the rechargeable pipette controller below:
https://www.sigmaaldrich.com/product/aldrich/z314951
Z315001
Hirschmann® pipetus® Junior pipette controller
for volumes 0.5 to 25 mL
Synonym(s):
Pipette controller
Select a Size
$358.00
Available to ship on22 April 2025Details
Select a Size
About This Item
$358.00
Available to ship on22 April 2025Details
Recommended Products
material
plastic
silicone
manufacturer/tradename
Hirschmann 9902000
volume
0.5-25 mL
General description
- Low cost, hand-powered model
- For volumes 0.5 to 25 mL
- Fine dosing valve for accurate adjustment of meniscus
- Discharge speed controlled by adjusting finger pressure on delivery trigger
- Silicone pipette holder accepts volumetric, serological or Pasteur pipettes
- Integral PTFE membrane filter prevents accidental ingress of liquid into controller
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
Is Hirschmann® pipetus® Junior pipette controller (catalog number Z315001) from Sigma-Aldrich rechargeable?
1 answer-
Helpful?
-