Skip to Content
Merck
All Photos(1)

Key Documents

51262-U

Ultra-High Performance Fingertight Fittings

PEEK (FingerTight nut (black) 10-32), for use with (1/16" tubing), pkg of 10 ea

Synonym(s):

High Pressure HPLC Fittings

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$173.00

$173.00


Available to ship on23 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 EA
$173.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$173.00


Available to ship on23 April 2025Details


Request a Bulk Order

material

PEEK (FingerTight nut (black) 10-32)

packaging

pkg of 10 ea

compatibility

for use with (1/16" tubing)

Looking for similar products? Visit Product Comparison Guide

General description

The innovative line of Upchurch Ultra High Performance fittings are desigined to withstand extreme temperature anf pressures. This line of fittings is perfect for use within the increasingly demanding requirements for today′s high performance analytical systems. The fittings are a 10/32 fingertight style for use with 1/16" and 1/32" OD tubing.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the backpressure rating of this fitting?

    1 answer
    1. The maximum pressure rating for this item is 12000 psi (827 bar). Please note that this is sufficient for conventional HPLC systems, but some UPLC methods exceed this pressure.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service