Skip to Content
Merck
All Photos(1)

Key Documents

Z227307

PEEK Tubing

L × O.D. × I.D. 10 ft × 1/16 in. × 0.005 in., red, max. pressure 8000

Synonym(s):

PEEK Tubing

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$123.00

$123.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$123.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32119290
NACRES:
SB.52

$123.00


Available to ship on22 April 2025Details


Request a Bulk Order

parameter

8000 max. pressure

L × O.D. × I.D.

10 ft × 1/16 in. × 0.005 in.

color

red

Looking for similar products? Visit Product Comparison Guide

General description

Polyetheretherketone (PEEK) tubing has become a popular replacement for stainless steel tubing in various places in the HPLC system. It is especially useful when contact between the sample and metal components must be avoided, such as when working with metal complexing agents or certain biochemical compounds. PEEK tubing also offers other advantages: it is flexible, easy to cut, has excellent mechanical stability and chemical compatibility, and can be used to 100 °C. Tetrahydrofuran (THF), dimethyl sulfoxide (DMSO), methylene chloride, and concentrated nitric and sulfuric acids should not be used with PEEK tubing. Our 1/16 in. PEEK tubing is color coded for easy identification of the internal diameter. 10 ft./3 m length.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service