Skip to Content
Merck
All Photos(1)

Key Documents

22668-U

Swagelok® Fittings Kit

Synonym(s):

TM=["Swagelok"] Connection Set

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$685.00

$685.00


Estimated to ship on18 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$685.00

About This Item

UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54

$685.00


Estimated to ship on18 April 2025Details


Request a Bulk Order

General description

A useful assortment of 1/8 in. and 1/4 in. brass fittings, packaged in a compartmented clear plastic storage box. Have the required parts on hand when needed, and save money (more than 30%, compared to purchasing items individually).

Application

Swagelok® fittings may be used in constructing a device mainly medium delivery system for steady-state growth of Methanococcus maripaludis in chemostats.[1] It may also be used in gas delivery chambers to prevent leaks of NH3.[2]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acute and prolonged effects of ammonia on hematological variables, stress responses, performance, and behavior of nursery pigs.
Von Borell, E., et al.
Journal of Swine Health and Production, 15, 137-137 (2007)
Andrew K Haydock et al.
FEMS microbiology letters, 238(1), 85-91 (2004-09-01)
To study global regulation in the methanogenic archaeon Methanococcus maripaludis, we devised a system for steady-state growth in chemostats. New Brunswick Bioflo 110 bioreactors were equipped with controlled delivery of hydrogen, nitrogen, carbon dioxide, hydrogen sulfide, and anaerobic medium. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service