Skip to Content
Merck
All Photos(1)

Key Documents

SML1814

Sigma-Aldrich

3,6-DMAD hydrochloride

≥98% (HPLC)

Synonym(s):

N9-(3-(Dimethylamino)propyl)-N3,N3,N6,N6-tetramethylacridine-3,6,9-triamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€108.00

€108.00


Check Cart for Availability


Select a Size

Change View
5 MG
€108.00

About This Item

Empirical Formula (Hill Notation):
C22H32N5 · xHCl
CAS Number:
Molecular Weight:
366.52 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€108.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow to orange

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C)CCCNC1=C2C(C=C(N(C)C)C=C2)=NC3=C1C=CC(N(C)C)=C3

InChI

1S/C22H31N5/c1-25(2)13-7-12-23-22-18-10-8-16(26(3)4)14-20(18)24-21-15-17(27(5)6)9-11-19(21)22/h8-11,14-15H,7,12-13H2,1-6H3,(H,23,24)

InChI key

UIWOQSJSDYDIQJ-UHFFFAOYSA-N

Biochem/physiol Actions

3,6-DMAD is also called as N9-(3-(dimethylamino)propyl)-N3,N3,N6,N6-tetramethylacridine-3,6,9-triamine. It prevents the development of multiple myeloma (MM) tumor xenografts.[1]
3,6-DMAD is an acridine derivative that selectively suppresses ER stress- (300 nM Thapsigargin) induced HT1080 cellular XBP1 mRNA splicing (Eff. conc. 500 nM), but not eIF2a phosphorylation, by directly inhibiting IRE1? RNase (endoribonuclease) activity and disrupting IRE1α oligomerization. 3,6-DMAD is shown to exhibit anti-multiple myeloma efficacy in cultures in vitro (%survival/[3,6-DMAD]/cell line/24 h = 13%/4 M/RPMI 8226 and 8%/1 μM/MM1.R) and completely suppress the expansion of established RPMI 8226 tumor in mice in vivo when administered via intraperitoneal injection (10 mg/kg q.o.d.).
3,6-DMAD is an acridine derivative.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dadi Jiang et al.
Molecular cancer therapeutics, 15(9), 2055-2065 (2016-06-17)
Using a luciferase reporter-based high-throughput chemical library screen and topological data analysis, we identified N-acridine-9-yl-N',N'-dimethylpropane-1,3-diamine (DAPA) as an inhibitor of the inositol requiring kinase 1α (IRE1α)-X-box binding protein-1 (XBP1) pathway of the unfolded protein response. We designed a collection of
Acridine Derivatives as Inhibitors of the IRE1 alpha-XBP1 Pathway Are Cytotoxic to Human Multiple Myeloma
Jiang D, et al.
Molecular Cancer Therapeutics (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service