Skip to Content
Merck
All Photos(1)

Documents

5.08316

Sigma-Aldrich

CTAP

Synonym(s):

CTAP, µ opioid receptor antagonist, CTAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H69N13O11S2
CAS Number:
Molecular Weight:
1104.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

solid

potency

3.5 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: soluble

storage temp.

−20°C

InChI

1S/C51H69N13O11S2/c1-26(65)39(42(53)68)62-49(75)41-51(3,4)77-76-25-38(61-43(69)33(52)21-28-11-6-5-7-12-28)47(73)59-36(22-29-16-18-31(67)19-17-29)45(71)60-37(23-30-24-57-34-14-9-8-13-32(30)34)46(72)58-35(15-10-20-56-50(54)55)44(70)63-40(27(2)66)48(74)64-41/h5-9,11-14,16-19,24,26-27,33,35-41,57,65-67H,10,15,20-23,25,52H2,1-4H3,(H2,53,68)(H,58,72)(H,59,73)(H,60,71)(H,61,69)(H,62,75)(H,63,70)(H,64,74)(H4,54,55,56)/t26-,27-,33-,35+,36+,37-,38+,39+,40+,41-/m1/s1

InChI key

OFMQLVRLOGHAJI-FGHAYEPSSA-N

General description

A highly potent octapeptide that acts as a selective and brain penetrating bioavailable µ opioid receptor antagonist (IC50 = 3.5 nM). Used in studies of reward systems and additions.

Biochem/physiol Actions

Primary Target
u Opioid receptpr

Warning

Toxicity: Standard Handling (A)

Sequence

D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH2 (2-7 disulfide bond)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Perry, C., et al., 2013. Behav. Neurosci.127, 535.
Burkett, J. et al. 2011. Neuropsychopharm.36, 2200.
Steinmiller, L., et al., 2008. Psychopharmacol. (Berl).195, 497.
Sterious, S., et al., 2003. J. Pharmacol. Exp. Ther.304, 301.
Abbruscato, T., et al. 1997. J. Pharmacol.280, 402.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service