Skip to Content
Merck
All Photos(1)

Key Documents

SML2045

Sigma-Aldrich

R-IMPP hydrochloride

≥98% (HPLC)

Synonym(s):

(R)-N-(isoquinolin-1-yl)-3-(4-methoxyphenyl)-N-(piperidin-3-yl)propanamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€89.30
25 MG
€291.00

€89.30


Estimated to ship on04 April 2025



Select a Size

Change View
5 MG
€89.30
25 MG
€291.00

About This Item

Empirical Formula (Hill Notation):
C24H27N3O2 · xHCl
CAS Number:
Molecular Weight:
389.49 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€89.30


Estimated to ship on04 April 2025


Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(CCC1=CC=C(OC)C=C1)N([C@H]2CNCCC2)C3=C(C=CC=C4)C4=CC=N3.[H]Cl

InChI

1S/C24H27N3O2.ClH/c1-29-21-11-8-18(9-12-21)10-13-23(28)27(20-6-4-15-25-17-20)24-22-7-3-2-5-19(22)14-16-26-24;/h2-3,5,7-9,11-12,14,16,20,25H,4,6,10,13,15,17H2,1H3;1H/t20-;/m1./s1

InChI key

OSIRDWNTKGVIOE-VEIFNGETSA-N

Biochem/physiol Actions

Potent and selective inhibitor of PCSK9 secretion
R-IMPP is a potent and selective inhibitor of proprotein convertase subtilisin/kexin type 9 (PCSK9) secretion that stimulates uptake of LDL-C in hepatoma cells by increasing LDL-R levels, without altering levels of secreted transferrin. R-IMPP binds selectively to human ribosomes and causes transcript-dependent inhibition of PCSK9 translation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donna N Petersen et al.
Cell chemical biology, 23(11), 1362-1371 (2016-10-18)
Proprotein convertase subtilisin/kexin type 9 (PCSK9) is a secreted protein that downregulates low-density lipoprotein (LDL) receptor (LDL-R) levels on the surface of hepatocytes, resulting in decreased clearance of LDL-cholesterol (LDL-C). Phenotypic screening of a small-molecule compound collection was used to identify an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service