Skip to Content
Merck

Skip To

5.08015

CTOP

Synonym(s):

CTOP, µ Opioid Receptor Antagonist, CTOP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 157,00

About This Item

Empirical Formula (Hill Notation):
C50H67N11O11S2
CAS Number:
Molecular Weight:
1062.26
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light

€ 157,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

Quality Segment

form

solid

potency

0.96 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

water: soluble

storage temp.

−20°C

SMILES string

S1SCC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C1(C)C)C(=O)NC(C(O)C)C(=O)N)C(O)C)CCCN)Cc4c5c([nH]c4)cccc5)Cc3ccc(cc3)O)NC(=O)C(N)Cc2ccccc2

InChI

1S/C50H67N11O11S2/c1-26(62)39(42(53)65)59-49(72)41-50(3,4)74-73-25-38(58-43(66)33(52)21-28-11-6-5-7-12-28)47(70)56-36(22-29-16-18-31(64)19-17-29)45(68)57-37(23-30-24-54-34-14-9-8-13-32(30)34)46(69)55-35(15-10-20-51)44(67)60-40(27(2)63)48(71)61-41/h5-9,11-14,16-19,24,26-27,33,35-41,54,62-64H,10,15,20-23,25,51-52H2,1-4H3,(H2,53,65)(H,55,69)(H,56,70)(H,57,68)(H,58,66)(H,59,72)(H,60,67)(H,61,71)

InChI key

PZWWYAHWHHNCHO-UHFFFAOYSA-N

General description

A potent antagonist highly selective for µ opioid receptor (Ki = 0.96 nM for µ receptor and >10,000 nM for δ receptors). Often used for studying the functional roles of µ opioid receptors in both central and peripheral nervous systems.

Biochem/physiol Actions

Primary Target
mu opioid

Physical form

Supplied as trifluoracetate salt

Other Notes

D-Phe-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH2 (Orn: ornithine; Pen: penicillamine; SS bond: 2-7)
Feng, Y., et al. 2012. Curr Drug Targets.13, 230.
Hawkins, N., et al. 1989. J. Pharmacol. Exp. Ther.248, 73.
Gulya, K., et al. 1988. Eur. J. Pharmacol.150, 355.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1408222P5296718165
assay

≥95% (HPLC)

assay

-

assay

≥97% (HPLC)

assay

97%

form

solid

form

buffered aqueous solution

form

powder

form

liquid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage condition

OK to freeze, protect from light

storage condition

-

storage condition

-

storage condition

-


Still not finding the right product?

Explore all of our products under CTOP


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters