Skip to Content
Merck

Skip To

SML0914

HA155

≥98% (HPLC)

Synonym(s):

(Z)-4-((4-((3-(4-Fluorobenzyl)-2,4-dioxo-1,3-thiazolan-5-yliden)-methyl)phenoxy)methyl)benzeneboronic acid, B-[4-[[4-[(Z)-[3-[(4-Fluorophenyl)methyl]-2,4-dioxo-5-thiazolidinylidene]methyl]phenoxy]methyl]phenyl]-boronic acid, HA 155

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 284,00

About This Item

Empirical Formula (Hill Notation):
C24H19BFNO5S
CAS Number:
Molecular Weight:
463.29
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
protect from light

€ 284,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

HA155 is a potent and selective autotaxin inhibitor, which potently blocks thrombin-induced lysophosphatidic acid (LPA) secretion in platelets.
HA155 is a potent and selective autotaxin inhibitor.

Other Notes

Light sensitive

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1491SML2328SML1915
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage condition

protect from light

storage condition

-

storage condition

desiccated

storage condition

-

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters