Skip to Content
Merck

Skip To

MAC142

Anti-Cas9 Antibody, N-terminal Antibody, clone 6H4 Antibody, S. aureus

clone 6H4, from mouse

Synonym(s):

SaCas9, CRISPR-associated endonuclease Cas-9

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
6H4, monoclonal
Application:
IF, IP, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6H4, monoclonal

species reactivity

Staphylococcus aureus, bacteria

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable, immunoprecipitation (IP): suitable, western blot: suitable

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABS2202AB356480MAC133
species reactivity

Staphylococcus aureus, bacteria

species reactivity

Streptococcus pyogenes, bacteria

species reactivity

Staphylococcus aureus, bacteria

species reactivity

-

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

serum

antibody form

purified antibody

clone

6H4, monoclonal

clone

polyclonal

clone

polyclonal

clone

7A9, monoclonal

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

wet ice

General description

CRISPR-associated endonuclease Cas9 (UniProt: J7RUA5; also known as SaCas9) is encoded by the Cas9 gene (Gene ID: in Staphylococcus aureus. CRISPR (clustered regularly interspaced short palindromic repeat) is an adaptive immune system that provides protection against mobile genetic elements (viruses, transposable elements and conjugative plasmids). CRISPR clusters contain spacers, sequences complementary to antecedent mobile elements, and target invading nucleic acids. CRISPR clusters are transcribed and processed into CRISPR RNA (crRNA). In type II CRISPR systems correct processing of pre-crRNA requires a trans-encoded small RNA (tracrRNA), endogenous ribonuclease 3 (rnc) and this protein. The tracrRNA serves as a guide for ribonuclease 3-aided processing of pre-crRNA. Subsequently Cas9/crRNA/tracrRNA endonucleolytically cleaves linear or circular dsDNA target complementary to the spacer.Cas9 is inactive in the absence of the 2 guide RNAs (gRNA). Cas9 recognizes the protospacer adjacent motif (PAM) in the CRISPR repeat sequences to help distinguish self-versus non-self, as targets within the bacterial CRISPR locus do not have PAMs. PAM recognition is also required for catalytic activity. Cas9 endonucleolytically cleaves linear or circular dsDNA target complementary to the spacer. The target strand not complementary to crRNA is first cut endonucleolytically, and then trimmed 3′-5′ exonucleolytically.
~124 kDa observed; 123.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: N-terminus
His-tagged recombiant Cas-9 from Staphylococcus aureus.

Application

Anti-Cas-9, N-terminal, clone 6H4, Cat. No. MAC142, is a mouse monoclonal antibody that detects Cas-9 in Staphylococcus aureus and its identity has been confimed by Isotyping Test and Western Blotting.
Research Category
Secondary & Control Antibodies
Western Blotting Analysis: 0.57 µg/mL from a representative lot detected CRISPR-associated endonuclease Cas9/Csn1 in whole cell lysates of HEK293 (WT), HEK293/Cas9 from S. aureus, or HeLa/Cas9 from S. pyogenes. (Courtesy of Stefan Schuchner, Ph.D. and Egon Ogris, M.D., Medical University of Vienna, Austria).

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected CRISPR-associated endonuclease Cas9/Csn1 in transiently transfected U20S cells expressing SaCas9-T2A-GFP. (Courtesy of Stefan Schuchner, Ph.D. and Egon Ogris, M.D., Medical University of Vienna, Austria).

Immunoprecipitation Analysis: 1-2 µg from a representative lot detected CRISPR-associated endonuclease Cas9/Csn1 in SaCas9. (Courtesy of Stefan Schuchner, Ph.D. and Egon Ogris, M.D., Medical University of Vienna, Austria).

Biochem/physiol Actions

Clone 6H4 detects Cas-9 in Staphylococcus aureus.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG1 .

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
MAC14204054839233135
MAC142-25UG04054839421051

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service