Skip to Content
Merck

Skip To

ABS2202

Anti-Cas9

from rabbit, purified by affinity chromatography

Synonym(s):

SpCas9, CRISPR-associated endonuclease Cas9

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

Streptococcus pyogenes, bacteria

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

158.44 kDa calculated.
CRISPR-associated endonuclease Cas9/Csn1 (UniProt: Q99ZW2; also known as SpCas9, SpyCas9) is encoded by the Cas9 (also known as Csn1) gene (Gene ID: 901176) in Streptococcus pyogenes. CRISPR (clustered regularly interspaced short palindromic repeat) is an adaptive immune system that provides protection against mobile genetic elements (viruses, transposable elements and conjugative plasmids). CRISPR clusters contain spacers, sequences complementary to antecedent mobile elements, and target invading nucleic acids. Cas9 has a bi-lobed architecture with a recognition lobe (residues 41-425) and a discontinuous nuclease lobe (NUC, residues 1-40 and 453-1053); the crRNA-target DNA lies in a channel between the 2 lobes. The NUC lobe has 2 endonuclease domains. The discontinuous RuvC-like domain in NUC cleaves the target DNA non-complementary to crRNA while the HNH nuclease domain cleaves the target DNA complementary to crRNA. Cas9 serves as a RNA-guided DNA endonuclease that cleaves double-stranded DNA targets with a protospacer adjacent motif (PAM) and complementarity to the guide RNA. The homologous region of the Staphylococcus aureus is not closely related in amino acid sequence in Streptococcus pyogenes, hence this antibody does not recognize that protein.

Immunogen

Epitope: C-terminus
KLH-conjugated linenar peptide corresponding to 18 amino acids from the C-terminal region of Streptococcus pyogenes.

Application

Anti-Cas9 Antibody, Cat. No. ABS2202, is a highly specific rabbit polyclonal antibody that targets Cas9 and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Cas9 in HEK293 cells transfected with Cas9 from Streptococcus pyogenes.

Immunocytochemistry Analysis: 1 µg/mL from a representative lot detected Cas9 in non-transfected HEK293H and HEK293H transfected with pCDNA3.0-Cas9.

Western Blotting Analysis: 10 µg/mL from a representative lot detected Cas9 in HEK293H cell lysate from cells transfected with pCDNA3.0-Cas9.
Research Category
Epigenetics & Nuclear Function

Biochem/physiol Actions

This rabbit polyclonal antibody detects Cas9 in Streptococcus pyogenes. It targets an epitope within 18 amino acids from the C-terminal region. This antibody does not react with Cas9 from Staphylococcus aureus.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HEK293 cells transfected with Cas9 from Streptococcus pyogenes.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Cas9 in HEK293 cells transfected with Cas9 from Streptococcus pyogenes.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB356480MABS1291MAC141
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

mouse

antibody form

affinity isolated antibody

antibody form

serum

antibody form

purified antibody

antibody form

purified antibody

UniProt accession no.

Q99ZW2

UniProt accession no.

J7RUA5

UniProt accession no.

Q99ZW2

UniProt accession no.

J7RUA5

clone

polyclonal

clone

polyclonal

clone

10C11-A12, monoclonal

clone

5F1, monoclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ABS220204054839355561
ABS2202-25UG04054839508455

Questions

Reviews

No rating value

Active Filters