Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 g | Check Cart for Availability | € 79,10 | |
| 5 g | Check Cart for Availability | € 243,00 |
About This Item
Quality Level
product line
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-30°C
General description
This pre-loaded Wang resin is prepared using a synthesis-tested base resin, to ensure reproducibility and efficacy in SPPS. Loading of the Fmoc-amino acid is carried out under conditions to minimize enantiomerization and dipeptide formation, the values of which are provided on the COA. The lot number of the batch of Fmoc-amino acid used to prepare the resin is also provided on the COA, to provide transparency and traceability.
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Analysis Note
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 1.00 mmol/g
Enantiomeric purity: ≥ 99.5 % (a/a)
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
Content Dipeptide (HPLC): ≤ 0.5 % (a/a)
Novabiochem Fmoc amino acid: lot number
The qualitiy of this resin has been confirmed by the successful synthesis of the 21mer peptide related to myelin oligodendrocyte glycoprotein (35-55), with an Fmoc-Val-Wang resin made from the same batch of base resin.
The polymer matrix is copoly (styrene - 1% DVB), 100 - 200 mesh.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis |
| product line Novabiochem® | product line Novabiochem® | product line Novabiochem® | product line Novabiochem® |
| functional group Fmoc | functional group Fmoc | functional group Fmoc | functional group Fmoc |
| Quality Level 400 | Quality Level 400 | Quality Level 400 | Quality Level 200 |
| application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis |
| storage temp. 2-30°C | storage temp. 2-30°C | storage temp. 2-30°C | storage temp. 2-30°C |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8560179999 | 04027269115807 |
| 8560170005 | 04027269115791 |
| 8560170001 | 04027269115784 |



