Skip to Content
Merck

Skip To

8.56016

Fmoc-Ser(tBu)-Wang resin (100-200 mesh)

Novabiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 85,70
5 g
Check Cart for Availability
€ 243,00
€ 182,25

About This Item

NACRES:
NA.22
UNSPSC Code:
12352209

€ 85,70


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

General description

Pre-loaded resin for synthesis of peptide acids containing a C-terminal serine amino-acid residue by Fmoc SPPS.
This pre-loaded Wang resin is prepared using a synthesis-tested base resin, to ensure reproducibility and efficacy in SPPS. Loading of the Fmoc-amino acid is carried out under conditions to minimize enantiomerization and dipeptide formation, the values of which are provided on the COA. The lot number of the batch of Fmoc-amino acid used to prepare the resin is also provided on the COA, to provide transparency and traceability.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 1.00 mmol/g
Enantiomeric purity: ≥ 99.5 % (a/a)
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
Content Dipeptide (HPLC): ≤ 0.5 % (a/a)
Novabiochem Fmoc amino acid: lot number
The qualitiy of this resin has been confirmed by the successful synthesis of the 21mer peptide related to myelin oligodendrocyte glycoprotein (35-55), with an Fmoc-Val-Wang resin made from the same batch of base resin.
The polymer matrix is copoly (styrene - 1% DVB), 100 - 200 mesh.

Other Notes

Replaces: 04-12-2062

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.560198.560218.56116
reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

Quality Level

400

Quality Level

400

Quality Level

200

Quality Level

400

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters