Skip to Content
Merck
All Photos(1)

Key Documents

8.56021

Sigma-Aldrich

Fmoc-Lys(Mtt)-Wang resin (100-200 mesh)

Novabiochem®

Synonym(s):

Fmoc-Lysine Mtt-Wang Resin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€121.00
5 G
€442.00

€121.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€121.00
5 G
€442.00

About This Item

UNSPSC Code:
12352209
NACRES:
NA.22

€121.00


Estimated to ship on22 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

General description

Pre-loaded resin for synthesis of peptide acids containing a side-chain modified C-terminal lysine amino-acid residue by Fmoc SPPS. The Mtt group can be removed selectively on the solid phase with 1% TFA in DCM, unmasking the side-chain lysine amino group for further functionalization.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Linkage

Replaces: 04-12-2067

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 1.00 mmol/g
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
The polymer matrix is copoly (styrene - 1% DVB) 100 -200 mesh.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service