Skip to Content
Merck

Skip To

07-2178

Anti-Abl interactor 1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Abl interactor 1, Abelson interactor 1, Abi-1, Abl-binding protein 4, AblBP4, Eps8 SH3 domain-binding protein, Eps8-binding protein, Nap1-binding protein, Nap1BP, Spectrin SH3 domain-binding protein 1, e3B1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 305,00

About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

€ 305,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

bovine ... Abi1(540815)
human ... ABI1(10006)
mouse ... Abi1(11308)
rat ... Abi1(79249)

General description

Abl interactor 1 (Abelson interactor 1) is a member of the family of Abl-interactors and is highly expressed in the brain. Abl interactor 1 is involved in the signaling of ABL1 and ABL2 nonreceptor tyrosine kinases, and the EGF-receptor. The Abl interactor 1 protein contains SH3 domains, and may function as an adaptor protein; It forms a trimeric complex with EPS8 and SOS1 in the transduction of signals from Ras to Rac. Previous studies have suggested that Abl interactor 1 via the Ras and Rac pathways may regulate cell proliferation, cell adhesion, and actin remodeling. Abl interactor 1 is also investigated for its role in tumor suppression.
~60 kDa observed. Uniprot describes many isoforms between 43-55 kDa that can be highly phosphorylated.

Immunogen

KLH-conjugated linear peptide corresponding to human Abl interactor 1.

Application

Detect Abl interactor 1 using this Anti-Abl interactor 1 Antibody validated for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate
Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Abl interactor 1 on 10 µg of Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
07-2129A510605-1938
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

14E5-G9, monoclonal

species reactivity

human, mouse

species reactivity

monkey, human

species reactivity

human, canine (predicted), mouse (predicted), rat

species reactivity

human

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

bovine ... Abi1(540815)
human ... ABI1(10006)
mouse ... Abi1(11308)
rat ... Abi1(79249)

Gene Information

human ... ABI1(10006)

Gene Information

human ... ABI1(10006)
mouse ... Abi1(11308)
rat ... Abi1(79249)

Gene Information

human ... G3BP1(10146)

UniProt accession no.

Q8IZP0

UniProt accession no.

Q8IZP0

UniProt accession no.

Q8IZP0

UniProt accession no.

Q13283


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters