Skip to Content
Merck
All Photos(1)

Documents

05-1938

Sigma-Aldrich

Anti-G3BP1 Antibody, clone 14E5-G9

clone 14E5-G9, from mouse

Synonym(s):

ATP-dependent DNA helicase VIII, GAP SH3 domain-binding protein 1, GAP binding protein, GTPase activating protein (SH3 domain) binding protein 1, Ras-GTPase-activating protein SH3-domain-binding protein, RasGAP-associated endoribonuclease G3BP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody product type

primary antibodies

clone

14E5-G9, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... G3BP1(10146)

General description

G3BP1 (Ras-GTPase activating protein SH3-domain binding protein 1) directly associates with SH3 domains of GTPase activating proteins (GAPs). It was first discovered by its binding to RasGAP. G3BPs have been shown to be involved in a number of mitogenic signaling pathways. G3BP1 is also known to be over expressed in many human cancers (Kim, 2007) and its expression appears to be inversely correlated with PTEN expression (Kim, 2007). Interestingly, G3BP1, along with G3BP2, has been shown to bind to the C-terminus of p53, playing a crucial role in its activity (Kim, 2007). Furthermore, the G3BPs have two traditional RNA binding motifs.

Specificity

This antibody recognizes G3BP1.

Immunogen

GST-tagged recombinant protein corresponding to human G3BP1.

Application

Anti-G3BP1 Antibody, clone 14E5-G9 is an antibody against G3BP1 for use in WB & IC.
Research Sub Category
RNA Metabolism & Binding Proteins

G-proteins

Quality

Evaluated by Western Blot in K562 cell lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected G3BP1 on K562 cell lysate.

Target description

52 kDa

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Sun et al.
Nature communications, 11(1), 3354-3354 (2020-07-06)
Expansion of an intronic (GGGGCC)n repeat region within the C9orf72 gene is a main cause of familial amyotrophic lateral sclerosis and frontotemporal dementia (c9ALS/FTD). A hallmark of c9ALS/FTD is the accumulation of misprocessed RNAs, which are often targets of cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service