07-2129
Anti-phospho-Abi-1 (Ser225) Antibody
from rabbit, purified by affinity chromatography
Synonym(s):
Abl interactor 1, Abelson interactor 1, Abi-1, Abl-binding protein 4, AblBP4, Eps8 SH3 domain-binding protein, Eps8-binding protein, Nap1-binding protein, Nap1BP, Spectrin SH3 domain-binding protein 1, e3B1
About This Item
biological source
rabbit
Quality Level
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
monkey, human
species reactivity (predicted by homology)
rat (based on 100% sequence homology), mouse (based on 100% sequence homology)
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
phosphorylation (pSer225)
Gene Information
human ... ABI1(10006)
General description
Specificity
Immunogen
Application
Cell Structure
Cytoskeletal Signaling
Quality
Western Blot Analysis: 1 µg/mL of this antibody detected Abi-1 in 10 µg of EGF-treated COS cells untreated and treated with lambda phosphatase.
Target description
55 kDa calculated.
Physical form
Storage and Stability
Analysis Note
EGF-treated COS cells untreated and treated with lambda phosphatase.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service