Skip to Content
Merck
All Photos(1)

Key Documents

DWK292612907

Duran® Connection Cap System Gl 45 With Cyan Pp Screw Cap

Synonym(s):

292612907, Connection Cap

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€365.00

€365.00


Check Cart for Availability


Select a Size

Change View
1 EACH
€365.00

About This Item

UNSPSC Code:
41123300
NACRES:
NB.17

€365.00


Check Cart for Availability

material

red PBT screw cap

feature

number of ports 4 (Stainless steel)

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 292612907

General description

The DURAN® 4-port connector cap is manufactured using an innovative additive manufacturing method called three-dimensional metal printing. This method allows the complex form to be optimised using computer-aided design software, as well as significantly reducing the amount of metal required for construction. The transfer connection cap can be used with all sizes of DURAN® GL 45 bottles, and facilitates the movement of liquids within a closed and aseptic system. The four male hose connectors are suitable for flexible tubing with a 6.4 mm (1/4") inner diameter. This corresponds to the SDD initiative Size A tubing type, allowing easy connection to buffer bags, mixing systems, etc. Freely rotating stainless steel centre allows the bottle to be unscrewed without the need to disconnect the tubing. Connectors and body are manufactured from Type 316L (1.4404 / S31603) stainless steel. A silicone gasket and the GL 45 screw cap provide a liquid tight seal. Wide temperature usage range (up to 180 °C). Autoclavable (121 °C for 15 minutes). Lot coded for full traceability and certification. Ideal for chemistry, life science and biopharma laboratories.

Application

small volumes of liquid need to be sampled or dispensed, such as a media supply for small bioreactors, in a perfusion circuit for cell culture, or oxygenation of very small samples

Features and Benefits

for 45 GL bottles Construction materials are certified as approved for food contact, and offer a high temperature resistance (up to 180 °C). The caps are fully autoclavable (121 °C / 15 minutes) and reusable

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service