Z742277
DURAN® replacement silicone gasket for the GL 45 stainless steel connection cap
diam. × thickness 40.5 mm × 3 mm, opening I.D. 28 mm
Synonym(s):
292232805
Select a Size
€41.40
Select a Size
About This Item
€41.40
Recommended Products
manufacturer/tradename
DURAN® 292232805
diam. × thickness
40.5 mm × 3 mm
opening I.D.
28 mm
Related Categories
General description
Features and Benefits
- Manufactured from stainless steel type 316L, PBT, and silicone
- Aseptic handling of liquids in pharmaceutical, biopharma and biotechnology applications: Easier attachment of flexible tubing by means of barbed connectors
- The one, two or three push-on tubing connectors on the upper and lower side are suitable for connecting flexible hoses with an internal diameter of 8.0 mm, such as Sani-Tech Ultra (ULTRA-C-312) silicone tubes
- The freely rotating stainless steel insert allows the bottle to be opened without disconnecting the tube connections
- Simplified transfer of liquids within closed and sterile systems through the bottle cap adapter
- All parts are fully welded, to prevent inclusions and cavities
- Hygienic, electropolished surface for efficient cleaning and sterilization
- Supplied complete with a USP <88> Class VI silicone O-ring gasket and a PBT GL 45 screw cap to provide a hygienic hermetic seal
- Lot coded for tracability and lot specific certification
- Replacement USP Class VI silicone O-ring gasket and PBT GL 45 screw open topped cap are available
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service